Learn More
Trajan Scientific and Medical Big Trap - Hydrocarbon
Product Code. 15730979
Click to view available options
Port Type:
1/4 in.
1/8 in.
Unit Size:
Each
Product Code. 15730979
Brand: Trajan Scientific and Medical 1035334
Only null left
Add to basket
Add to basket
For bulk purification applications or where several instruments are plumbed from a single source, a Big Trap gas filter is an ideal solution. A one-piece heavy walled alminium tube provides 750cm3 of capacity and a pressure rating of up to 250 psig
  • High capacity filters ideal for bulk purification - capacity of 80 g organics, depending upon impurities
  • Efficiency < 15 ppb
  • Can be used where several instruments are plumbed from a single gas source
  • Sintered stainless steel frits prevent particulate contamination
Specifications
For Use With (Application) FID for air and hydrogen, PID when used with an oxygen filter
Model Hydrocarbon
Volume (Metric) 750 cm3
Port Type 1/4 in.
Description Removes organic compounds from gas streams
For Use With Big Trap Mounting Clip
Pressure 250 psig