Learn More
Keystone Cleanroom Products BHD Autoclavable Wrapper
BIOPHARMAGREENER_CHOICE
Product Code. p-7010013 Shop All Keystone Cleanroom Products Products
Click to view available options
Length (English):
1.5 in.
2 in.
3 in.
4 in.
6 in.
Width (English):
1.5 in.
2 in.
3 in.
4 in.
6 in.
Unit Size:
Pack of 3000
Product Code. 17798177
Brand: Keystone Cleanroom Products BHDSHEET01.5X01.5
Only null left
Add to basket
Add to basket
When secured with autoclave tape or elasticized bands, these BHD Autoclavable Wrappers provide an adequate microbial barrier from external sources of contamination.
  • Designed for steam sterilization
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient microbial barrier
  • Resists tearing and punctures
  • Does not contain any natural latex rubber
  • Processed and packaged in ISO Certified Cleanrooms
  • Certificate of Conformance included with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance
Closure Type None
Color Blue
Disposable Single-use
For Use With (Application) Gamma, EtO, or Steam Sterilization
Material Cellulose
Sterility Non-sterile
Latex Free Yes
Temperature Range (Metric) Up to 135°C
Length (English) 1.5 in.
Length (Metric) 3.8 cm
Width (English) 1.5 in.
Width (Metric) 3.8 cm
Product Type Sterilization Wrap (Heavy-duty)