Learn More
Gibco™ BG-11 Media, optimized for cyanobacteria
BG-11 Media, optimized for cyanobacteria
Brand: Gibco™ A1379901
Description
Gibco™ BG-11 Media is optimized for the growth and maintenance of select Cyanobacteria including Synechococcus elongates. The formulation comes ready-to-use at 1X to let you avoid laborious media preparation steps.
Each Gibco™ BG-11 Media comes in the award winning Gibco™ bottle designed for easier use in the biosafety cabinet, minimizing the risk of contamination, and helps you perform cell culture more consistently. Together, the superior packaging and quality, greater reliability, and improved consistency in Cyanobacteria culture, results in better overall efficiency and more robust data (Fig. 1, 2).
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
Specifications
Gibco™ BG-11 Growth Media contains: • 1 L Gibco™ BG-11 Media — Store at 4 °C |
|
Algal Media | |
Microbiology Media | |
1 L |
Liquid | |
None (Ready-to-Use) | |
Cyanobacteria | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2526568 | Certificate of Analysis | 29/01/2025 | A1379901 |
2526566 | Certificate of Analysis | 29/01/2025 | A1379901 |
2824774 | Certificate of Analysis | 28/01/2025 | A1379901 |
2824778 | Certificate of Analysis | 27/01/2025 | A1379901 |
2824776 | Certificate of Analysis | 26/01/2025 | A1379901 |