Learn More
Fisherbrand™ Radiation Safety Beta Bins
Shop All Fisherbrand Products
Click to view available options
Capacity:
122 L
Unit Size:
Each
Description
- Radiation Safety Large Beta Bins With Wheels
- Beta – 122L with wheels – (74 x 41 x 41cm), use with bag CSR-BAG6
- Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
- Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
- Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
- Optional heavy duty drawstring bags may also be purchased
Specifications
Specifications
| For Use With (Equipment) | Accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes |
| Height (Metric) | 760mm |
| Product Type | Radiation Bin |
| Width (Metric) | 426mm |
| Capacity | 122 L |