Learn More
Bestatin hydrochloride, 98%, Thermo Scientific Chemicals
Product Code. 15453169
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Product Code. 15453169
Brand: Thermo Scientific Alfa Aesar J60384.MB
Request Bulk or Custom Format
Bestatin is an inhibitor of amino peptidases and a potent, irreversible inhibitor of LTA4 hydrolase. It inhibits the amino peptidase activity of LTA4 hydrolase with a Ki value of 201 nM. It offers promise as a novel analgesic because it protects endogenous opioid peptides against degradation. It does not inhibit carboxypeptidases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bestatin is an inhibitor of amino peptidases and a potent, irreversible inhibitor of LTA4 hydrolase. It inhibits the amino peptidase activity of LTA4 hydrolase with a Ki value of 201 nM. It offers promise as a novel analgesic because it protects endogenous opioid peptides against degradation. It does not inhibit carboxypeptidases.
Solubility
Soluble in water (4 mg/ml, warm), DMSO (>25 mg/ml), methanol, acetic acid, dimethyl formamide, and 100% ethanol.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 65391-42-6
Molecular Formula C16H25ClN2O4
Molecular Weight (g/mol) 344.836
MDL Number MFCD00058004
InChI Key XGDFITZJGKUSDK-UDYGKFQRSA-N
Synonym bestatin hydrochloride, n-2s,3r-3-amino-2-hydroxy-4-phenylbutyryl-l-leucine hydrochloride, ubenimex hydrochloride, bestatin hcl, bestatin monohydrochloride, c16h24n2o4.hcl, bestatin hydrochloride hplc, bestatin, 2s,3r-3-amino-2-hydroxy-4-phenylbutanoyl-l-leucine hydrochloride
PubChem CID 11957481
IUPAC Name (2S)-2-[[(2S,3R)-3-amino-2-hydroxy-4-phenylbutanoyl]amino]-4-methylpentanoic acid;hydrochloride
SMILES CC(C)CC(C(=O)O)NC(=O)C(C(CC1=CC=CC=C1)N)O.Cl
Specifications
Melting Point 216°C to 218°C
Color White
Quantity 25 mg
Beilstein 4628066
Solubility Information Soluble in water (4mg/ml,warm),DMSO (>25mg/ml),methanol,acetic acid,dimethyl formamide,and 100% ethanol.
Formula Weight 344.87
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bestatin hydrochloride