missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Besifloxacin hydrochloride

Product Code. 17221206
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 405165-61-9
Molecular Formula C19H22Cl2FN3O3

Specifications

Quantity 100 mg
Formula Weight 430.31
InChI Key PMQBICKXAAKXAY-UHFFFAOYNA-N
SMILES Cl.NC1CCCCN(C1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1Cl)C(O)=O
IUPAC Name 7-(3-aminoazepan-1-yl)-8-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
Molecular Weight (g/mol) 430.30
Chemical Name or Material Besifloxacin hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.