missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl alcohol, Puriss. p.a., ACS reagent, ≥99.0% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
4 x 25 mL
500 mL
2.5 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
CAS Max % | ≤100.0000% |
Melting Point | -16°C to -13°C (literature) |
Density | 1.045 g/mL at 25 °C(lit.) |
Boiling Point | 205.3°C |
Flash Point | 100.4°C |
Packaging | Glass Bottle |
Linear Formula | C6H5CH2OH |
Quantity | 4 x 25 mL |
Beilstein | 878307 |
Formula Weight | 108.14 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'