missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl fluoride, 97%

Product Code. p-7077261 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10612952

Brand: Thermo Scientific Acros 402040050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 455-32-3 | C7H5FO | 124.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-32-3
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.11
MDL Number MFCD00000364
InChI Key HPMLGNIUXVXALD-UHFFFAOYSA-N
Synonym benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa
PubChem CID 67999
IUPAC Name benzoyl fluoride
SMILES FC(=O)C1=CC=CC=C1

Specifications

Melting Point -28.0°C
Density 1.1500g/mL
Boiling Point 159.0°C to 161.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5COF
Refractive Index 1.496
Quantity 5 g
Beilstein 09,181
Specific Gravity 1.15
Solubility Information Solubility in water: hydrolyses.
Formula Weight 124.11
Percent Purity 97%
Chemical Name or Material Benzoyl fluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzoyl fluoride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P240-Ground/bond container and receiving equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P390-Absorb spillage to prevent material damage.
  • P402-Store in a dry place.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.