Learn More
Product Code | Brand | Description | Length (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Length (English) | Price | Quantity & Availability | |||||
11800830
|
Thermo Scientific™
PH479X1 |
Hearth Tray | 6 in. |
€ 94.37
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
|||||
11806695
|
Thermo Scientific™
PH480X1 |
Hearth Tray | 10 in. |
€ 70.85
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
11852585
|
Thermo Scientific™
SH480X1 |
Ceramic shelf | 6.87 in. |
€ 96.18
Each |
Estimated Shipment: 11-08-2025
Log in to see stock. |
|||||
Description
Use Thermo Scientific™ Benchtop Muffle Furnaces Accessories with Thermo Scientific™ Thermolyne™ Benchtop Muffle Furnaces.
Product Title
Thermo Scientific™ Benchtop Muffle Furnaces Accessories