Learn More
Bel-Art™ SP Scienceware™ Vacuum Pressure Indicator
Product Code. 11824792 Shop All SP Scienceware Products
Click to view available options
For Use With (Equipment):
Fits on to tapered end of stopcock
Unit Size:
Each
Product Code. 11824792
Brand: Bel-Art™ H420420000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Indicates if desiccator is under vacuum.
  • Thermoplastic elastomer collapses with negative vacuum pressure in desiccator and expands when vacuum is absent
  • Fits onto tapered end of stopcock
Specifications
Product Type Vacuum Pressure Indicator
For Use With (Application) Thermoplastic elastomer bellows collapses when under negative pressure (vacuum), expands when under ambient atmospheric pressure, indicates the desiccator vacuum
For Use With (Equipment) Fits on to tapered end of stopcock