Learn More
Bel-Art™ Techni-Dome™ 360 Gas-Ported Desiccator
Shop All SP Scienceware Products
Click to view available options
Volume (Metric):
65 L
Unit Size:
Each
Description
- Protects generous quantities of standards, samples, electronics or other moisture-sensitive items
- Spacious 65 liter (4000 cu. in.) internal volume can hold items up to 45 cm (18 in.) tall or wide, including racks, trays and instruments
- Two gas ports are tapered and accept tubing with 0.25 to 0.375 in. I.D.; easy turning PTFE stopcocks facilitate turbulence-free entrance of gases, minimizing disturbance of contents
- Polycarbonate construction is resistant to most common chemicals, as well as thermal stress; wipes clean easily
- Includes two removable, polypropylene, 36.5cm (14.5 in.) and 22.5 cm (9 in.) diameter shelves
- Side handles provide easy portability and can be used as a place to affix tamper-evident seals
- Gas ports can be closed to use desiccator with standard desiccant cartridges (08-594-17C, 08-594-17B, or 08-594-14B)
Specifications
Specifications
| Color | Transparent |
| Material | Polycarbonate |
| Product Type | Gas-ported Desiccator |
| Includes | 2 gas ports, 2 removable polypropylene shelves (14.5 in. and 9 in.), polypropylene stopcock with PTFE plug and red handle |
| Volume (Metric) | 65 L |
| Height (Metric) | 45 cm |
| Height (English) | 18 in. |
| Diameter (Metric) Inner | 36.5 cm |
| Diameter (Metric) Outer | 45 cm |
| Weight (English) | 11 lb. |
| Show More |