missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ ProCulture Stak-A-Tray™ System

Maximize incubator, cold room and bench space.

Brand:  Bel-Art™ H18610-0420

 View more versions of this product

Product Code. 17672215

  • € 690.00 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Keeps trays, flasks and dishes level and stable for even settling of tissue culture cells
  • Racks interlock when stacked, to a maximum of 5 racks high (allows up to 20 levels of trays)
  • Each rack tier holds one large tray (Mfr. No. 18610-1440), 35.6 x 35.6cm, or two smaller trays (Mfr. No. 18610-0470), 17.8 x 35.6cm
  • Perforated trays allow air, gas and water vapor circulation
  • Racks and trays sold separately
  • Stainless-steel storage system is autoclavable

Rack, 2-Tier (Mfr. No. 18610-0420)

  • 5cm clearance between trays, ideal for storing larger culture flasks
  • Rack, 4-Tier (Mfr. No. 18610-0440)
    • 2.4cm clearance between trays, for storing Petri dishes or smaller culture flasks

    Specifications

    Specifications

    Rack Frame: 2 Tier Rack
    Stainless Steel
    37.5 cm
    35.8 cm
    10.8 cm
    Autoclavable
    37.5 x 35.8 x 10.8cm (14.75 x 14.0625 x 4.25 in.)
    14.75 in.
    14.0625 in.
    4.25 in.
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers