Learn More
Bel-Art™ Mini Sampler Spoons
Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
0.05 mL
0.1 mL
0.25 mL
0.5 mL
1 mL
Unit Size:
Each
Pack of 25
Description
- These disposable mini measures are designed for retrieving small, specific volume samples of powders or similar materials
- Made from white high-impact polystyrene plastic
- All sterile products are lot stamped for accurate tracing
Specifications
Specifications
| Sterility | Sterile |
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 0.05 mL |
| Color | White |
| Disposable | Single-use |
| Product Type | Spoon |
| For Use With (Application) | Designed for retrieving small, specific volume samples of powders or similar materials |
| Material | Polystyrene |