missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ ProCulture™ Petri Dish Racks

Increase workspace and efficiently transport large quantity of Petri dishes

Brand:  Bel-Art™ F18982-0309

Product Code. 11774736

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Made of lightweight, chemically resistant polycarbonate and autoclavable at 121°C (250°F), these racks are perfect for use in an incubator
  • Silicone holding strips on the rack's four columns grip each Petri dish and enable the user to easily insert and remove groups of dishes without disturbing upper or lower dishes
  • Labels on the sides of the rack provide space for critical sample identification
  • Dimensions: 24.1cm (9.5 in.) high
Specifications

Specifications

Petri Dish Rack
Autoclavable
Petri Dishes
48
No
10cm Petri dishes
184 mm
9 in.
12 x 4
Blue
Polycarbonate
Rectangular, Rounded Corners
Non-sterile
184 mm
22.86 cm
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ ProCulture™ Petri Dish Racks > Holds forty-eight 100mm petri dishes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.