Learn More
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector
Shop All SP Scienceware Products
Click to view available options
For Use With:
4.7 to 6mm
4.7 to 9.5mm
6 to 12.7mm
9.5 to 12.7mm
Fits 9 x 6mm Tubing I.D.
Unit Size:
Pack of 12
Description
- Chemical-resistant polypropylene plastic for use with wide range of liquids or gases
- FDA-grade for direct contact with food, drugs, and cosmetics
- Autoclavable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | 4.7 to 9.5mm |
| Tubing Size | For 3/16 in. to 1/2 in. tubing (4.7 to 9.5mm) |
| Includes | Reducing connector |
| Color | Clear |
| Product Type | Barbed Fitting |