Learn More
behr Labor-Technik™ behropur™ Water Deionizer Made Of Nylon
Product Code. p-7217110
Click to view available options
Flow Rate:
300 L/h
500 L/h
500 L/hr.
800 L/h
Capacity:
1200 L
2400 L
5500 L
Unit Size:
Each
Product Code. 10544862
Brand: behr Labor-Technik™ B00223475
Only null left
Add to basket
Add to basket
Made from Nylon. Convenient and safe mixed-bed ion exchanger for small to medium-sized quantities of high purity water. To supply laboratory glassware washers, for general laboratory maintenance and for small industrial consumers. For subsequent demineralization of reverse osmosis systems.
  • Optimum exploitation of the ion exchanger capacity due to absolute uniform water distribution in the unit
    • Can be connected to the raw water network directly and without a pressure reducer
      • Resistant to alternating pressure
        • Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
          • Also available with shutoff when the limited value is reached and limit control
            • Pressure resistant to 8 bar
Specifications
Description B10DNZ Pressure resistant mixed-bed unit made of nylon, permanent pressure resistant to 8 bar, spare cartridge
Capacity 1200 L
Flow Rate 300 L/h
Material Nylon
Model B10DNZ
Type Demineralizers
Width (Metric) 22 cm
Height (Metric) 22 cm
Depth (Metric) 60 cm