Learn More
Beclin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11306-1-AP-20UL
 View more versions of this product
Product Code. 16828293
  • € 145.00 / 20µL
Description
Beclin 1 plays a central role in autophagy. It acts as a core subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate. Different PI3K complex forms are believed to play a role in multiple membrane trafficking pathways - PI3KC3-C1 is involved in the initation of autophagosomes and PI3KC3-C2 is involved in the maturation of autophagosomes and endocytosis. Beclin 1 also plays a role in antiviral host defense.
Specifications
Specifications
Beclin 1
Polyclonal
Unconjugated
BECN1
ATG6, Beclin 1, beclin 1, autophagy related, beclin1, BECN1, GT197, Protein GT197, VPS30
Rabbit
Antigen Affinity Chromatography
RUO
114558, 56208, 8678
-20°C
Liquid
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O88597, Q14457, Q91XJ1
Becn1
Beclin 1 Fusion Protein Ag1843
20 μL
Primary
Human, Rat, Mouse