missing translation for 'onlineSavingsMsg'
Learn More

Terracycle™ Beard Nets and Hair Nets Zero Waste Box™
GREENER_CHOICE

Product Code. 17875725
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17875725

Brand: Terracycle™ CS43MNL

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Recycle Beard Nets and Hair Nets using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

Use this Zero Waste Box to recycle:

  • Beard nets
  • Hair nets

Approximate waste units accepted:

  • Small: 1000 units
  • Medium: 2700 units
  • Large: 5000 units

Do not send in:

  • Medical waste
    • This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
  • Disposable masks or gloves
  • Lab coats or other garments
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
  • Batteries, pressurized canisters, broken glass and medical waste

Specifications

Capacity (Metric) 15 kg
Capacity (English) 33 lb.
Color Beige, Green, Red, White
Dimensions (L x W x H) 30.5 x 30.5 x 84 cm
Disposable Single-use
Latex Free Yes
Size Medium
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.