Learn More
Fisherbrand™ Bead Mill 4 Adapter Kit
An adapter for bead mill homogenizers to expand their capacity.
Brand:  Fisherbrand™ 15555809
Product Code. 15555809
  • € 202.00 / Each
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 2mL and 7mL
  • Aids in homogenization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapter
2 mL and 7 mL
Videos
  • Fisherbrand bead mills and bead kits
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand bead mills and bead kits
    SDS
    Documents
    Documents
    Flyers
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Fisherbrand™ Bead Mill 4 Adapter Kit >