Learn More
BD Luer-Lok™ Access Device
Use for needleless collection of blood. BD Vacutainer™ Luer-Lok™ Access Device is a direct access collection device.
Brand:  BD 36490200
Product Code. 10782337
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For needleless collection of blood
  • Holder with preattached multiple sample, male adapter
  • Compatible with female luer connections or IV ports designed for luer access
  • Allows specimens to be collected into an evacuated tube
  • Has blue color-coded threads to provide easy differentiation from other holder-based products
  • Provides the security of a threaded locking luer connection, replacing luer slip devices
  • Compatible with luer-activated ports for closed, direct urine sampling found in some Foley catheters (such as the Bard™ E-Z™ Lock Sampling Port)
  • Sterile; latex-free
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Male Luer
Luer activated ports for closed, direct Urine sampling found in some Foley catheters
Blood Collection Access Device
For needle-less collection of Blood
Yes
Sterile
Certificates
Special Offers
Special Offers
Non-returnable medical device