Learn More
BD Biotin Human CD8 T Lymphocyte Enrichment Set - DM
For the negative selection of CD8 T lymphocytes from peripheral blood
Brand:  BD 557941
1305.13 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15809128
  • € 1310.00 / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Description
  • Avoids inadvertent activation of the enriched CD8 T lymphocytes
Magnetic Labeling
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Biotin
1 x 109 Cells
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Biotin Human CD8 T Lymphocyte Enrichment Set - DM > 5mL