missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Soluble Starch

Product Code. 11768253
Click to view available options
:
Soluble Starch; 500g
This item is not returnable. View return policy

Product Code. 11768253

Brand: BD 217820

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides Starch for hydrolysis, detoxification of metabolic byproducts and as a carbon source

  • Improves growth response
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2 to 25°C
Additive Soluble Starch
Form Powder
For Use With (Application) Provides Starch for hydrolysis, detoxification of metabolic byproducts and as a carbon source
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.