Learn More
APC BrdU Flow Kit, 200 tests, BD
missing translation for 'orderingAttributeHoverText'
Quantity:
4 x 50 Tests
missing translation for 'unitSize'
1 set
Beschreibung
Povides reagents for immunofluorescent BrdU staining with a protocol, compatible with the use of additional fluorescent antibodies specific for other cellular molecules. Used with fluorescent anti-cytokine antibodies in analyses of cultured cells following in vitro mitogenic stimulation of quiescent lymphoid cell populations.
- Consists of Flow cytometer with 488nm laser capable of detecting 7-AAD and a 633- to 640nm laser capable of detecting APC
- Staining procedure includes the fixative paraformaldehyde
Spezifikation
Spezifikation
| Content And Storage | 4°C |
| Includes | One 65μL vial of APC-conjugated Anti-BrdU antibody, one 25mL bottle Cytofix/Cytoperm™ buffer, two 25mL bottles perm/Wash™ buffer (10X), one 10mL bottle Cytoperm™ |
| No. of Tests | 200 |
| Quantity | 4 x 50 Tests |
| Validated Application | RUO |
| Product Type | APC BrdU Flow Kit |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur