Learn More
BINDER™ Base
Base for use with C 170 and CB 170.
Brand: BINDER™ 9051-0028
This item is not returnable.
View return policy
Description
- For use with CB 170
- Includes casters
Specifications
Base - the base equipped with casters is used for the secure positioning and leveling of a BINDER CO2 incubator | |
Steel | |
Casters |
BINDER Chambers CB-S 170, CB 170, CBF 170 | |
Stand |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
BINDER™ Base > Base; Includes: Casters