Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Nanopure II, Older Nanopure Systems
Nanopure Infinity Systems
Nanopure Quad-style System
Feedwater Source:
Deionized, Tap
Pretreated
Reverse Osmosis, Distilled
Tap
Tap or Pretreated
Model:
D3804
D3805
D4801
D4802
D50254
D50255
D50256
D50257
D5026
Unit Size:
Each
Description
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Specifications
Specifications
| Feedwater Source | Deionized, Tap |
| For Use With (Application) | Water purification cartridge |
| For Use With (Equipment) | Nanopure Infinity Systems |
| Includes | One 09-050-226 Tap/DI Pretreatment, three 09-050-247 Ultrapure™ cartridges and two 09-050-279 Remote Dispenser 0.2μm filters |
| Model | D50256 |
| Type | Cartridge Kit |