Learn More
Hamilton™ Barbed Fitting
Product Code. 10690834
Click to view available options
Diameter (Metric) Inner:
1.5 mm
Unit Size:
Each
Product Code. 10690834
Brand: Hamilton™ 35032
Only null left
Add to basket
Add to basket
For ease of connections in the laboratory.
  • Manufactured from CTFE
  • 87 psi/6 bar maximum pressure
  • Provided individually
Specifications
Type Barbed Fitting
Diameter (Metric) Inner 1.5 mm