Learn More
Banoxantrone-d12 dihydrochloride, MedChemExpress
MedChemExpress Banoxantrone-d12 (AQ4N-d12) dihydrochloride is the deuterium labeled Banoxantrone dihydrochloride. Banoxantrone is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.
Brand: Medchem Express HY-13562AS
This item is not returnable.
View return policy
Description
Banoxantrone-d12 (AQ4N-d12) dihydrochloride is the deuterium labeled Banoxantrone dihydrochloride. Banoxantrone is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.Specifications
Banoxantrone-d12 dihydrochloride | |
1 mg | |
Research | |
AQ4N-d12 dihydrochloride | |
529.48 | |
98.02% | |
Cancer-programmed cell death |
1562066-98-1 | |
Blue | |
C22H18D12Cl2N4O6 | |
O=N(C([2H])([2H])[2H])(C([2H])([2H])[2H])CCNC1=CC=C(NCCN(C([2H])([2H])[2H])(C([2H])([2H])[2H])=O)C(C(C2=C(O)C=CC(O)=C23)=O)=C1C3=O.Cl.Cl | |
529.48 | |
Research | |
Solid |
Research purposes only