missing translation for 'onlineSavingsMsg'
Learn More

Schneider Gerd™ Bandage Scissors

€ 8.50 - € 11.00

Specifications

Product Type Bandage Scissors
View More Specs

Products 3
Product Code Brand Tip Style Length (Metric) Price Quantity & Availability  
Product Code Brand Tip Style Length (Metric) Price Quantity & Availability  
10346584
View Documents Promotion Details
Schneider Gerd™
1015
Sharp-Blunt 160 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10450765
View Documents Promotion Details
Schneider Gerd™
1021-000
Blunt 160 mm
€ 11.00
Each
Estimated Shipment: 18-11-2024
to see stock.
Add to basket
 
10653395
View Documents Promotion Details
Schneider Gerd™
1013
Sharp/Blunt 130 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

Bandage Scissors
Videos
SDS
Documents

Documents

Product Certifications