Learn More
behr Labor-Technik™ Ballon Canister
The blue behroplast™ canisters are light fast. Your level display made of polyurethane ensures additional usage options. Naturally, they also have the welded tap for maximum tightness and optimum usage value.
Brand:  behr Labor-Technik™ B00176996
Product Code. 10024710
  • € 222.00 / Each
Explore more special offers
Description
Description
  • Canister
  • blue with light protection
  • level indicator and tap
  • Simple transport of the behrotest™ canister
  • Space-saving injected handle
  • Easy removal of liquids
  • Very easy removal with the welded behrotest™tap
  • Practical dimensions
  • Space-saving storage
  • Level display made of polyurethane
  • The liquid level directly in view
  • Individually packed canisters
  • Protection against damage and soiling
Specifications
Specifications
Ballon Canister
Blue
Tap and Level Indicator
250 mm
20 L
LDPE
280 mm
370 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers