Learn More
BICASA Balance Table (Vibration-proof)
Vibration-proof Balance Table with HPL countertop and marble plate.
Brand:  BICASA BT.0900.890
1604.68 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Product Code. 17417363
  • € 1850.00 / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Description
  • Specifically designed and manufactured to use with balances and analytical instruments
  • Marble vibration-proof central plate (Carrara White), Dimensions (L x W x Thickness): 520 x 350 x 40 mm
  • Fire-retardant particleboard filler body
  • HPL countertop
  • Light Gray color (2 decorative)
  • Separate internal welded tubular steel frame
  • With levelling feet and CAU devices