missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bode Chemie™ Baktolan™ Cleansing Lotion
Keep skin smooth and pliable even with intensive and repeated daily use with Bode Chemie™ Baktolan™ Cleansing Lotion with Allantoin and wheat protein. The pH value of 5.5 preserves the sensitive protective acid mantle of the skin.
Brand: Bode Chemie™ 9813333
Description
- Shelf Life:12 Months after opening
- pH Range: approximately 5.5
- Quantity: 1Pack
- Fragrance: Yes
Specifications
Sensitive Cleansing Lotion | |
Approximately 5.5. | |
500 mL |
Bottle | |
1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction