Learn More
Bode Chemie™ Baktolan™ Cleansing Lotion
Keep skin smooth and pliable even with intensive and repeated daily use with Bode Chemie™ Baktolan™ Cleansing Lotion with Allantoin and wheat protein. The pH value of 5.5 preserves the sensitive protective acid mantle of the skin.
Brand: Bode Chemie™ 9811373
Description
- Water-in-oil-in-water emulsion
- Protects against aqueous solutions and regenerates cracked, heavily stressed skin
- Innovative recipe (W / O / W emulsion)
- Protection against water-soluble substances
- Support of skin regeneration
- Strengthening the skin barrier
- Very good skin tolerance
- Free of perfume, silicone and dyes
- Counteracts occlusion-related irritation under the glove
Specifications
Cleansing Lotion | |
Approximately 5.5 | |
100 mL |
Tube | |
1 Ea. |