Learn More
Novacyt - Microgen Bioproducts Bacterial ID System Reagent/Consumable
Shop All Novacyt Group Products
Click to view available options
Product Type:
Indole Kovacs Reagent
Mineral Oil
Ninhydrin-filled Vial
Nitrate A Reagent
Nitrate B Reagent
Oxidase Strips
PYR Reagent
TDA Reagent
VP I Reagent
VP II Reagent
Unit Size:
10mL
50 strips
50mL
Description
- Replacement/supplemental reagent/consumable.
- From a range of simple, reliable, bacterial identification systems for food, environmental, and industrial laboratories.
- These tests will arm professionals with the necessary information to maximize the effectiveness of appropriate patient management.
- The range is expanding quickly and will encompass the widest range of microbiological testing solutions.
Specifications
Specifications
| Product Type | TDA Reagent |
| For Use With (Application) | Biochemical Identification Kits |
Safety and Handling
Product Identifier
- TDA REAGENT
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- MIXTURE LIST-Contains : Iron (III)Chloride,Hydrochloric acid