Learn More
Bode Chemie™ Bacillol™ Tissues
Bacillol™ Tissues - Alcohol-based rapid disinfectant wipes in a practical dispenser.
Brand: Bode Chemie™ 9813114
This item is not returnable.
View return policy
Description
- Ready-to-use disinfectant wipes
- Quickly and comprehensively effective
- Broad material compatibility with alcohol-resistant surfaces
- Aldehyde, dye and perfume free
- Easy removal from practical dispenser box
- Refillable
- Required spectrum of activity SARS-CoV-2: limited virucidal activity
Specifications
Disinfectant Wipes | |
Single-use | |
80 Wipes |
Pop-up canister | |
Disinfection of Alcohol-Resistant Surfaces and Medical Devices |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction
Product Title
Bode Chemie™ Bacillol™ Tissues > Tissues; Container Type: Flow Pack