missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synergy Healthcare™ Azowipe™ Hard Surface Disinfecting Wipes
Provides rapid disinfection of hard surfaces. Synergy Healthcare™ Azowipe™ Hard Surface Disinfectant Wipes are non-smearing, strong and durable for everyday use.
Brand: Synergy Healthcare™ 00723
This item is not returnable.
View return policy
Description
- 70% Isopropyl alcohol disinfectant
- Broad spectrum activity against a range of micro-organisms
- Extremely fast contact times from 15 seconds
- Non-smearing and streak-free when used on hard surfaces.
- Suitable for food manufacturing, hospitality and catering sectors.
- Compliant with HTM-01-05 guidelines
Specifications
Disinfectant Wipe | |
20 x 22 cm | |
200 Wipes/Dispenser |
Single-use | |
Wiper | |
70% isopropyl alcohol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- AZOWIPE Equipment Wipes
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Synergy Healthcare™ Azowipe™ Hard Surface Disinfecting Wipes > Size: 20 x 22cm
Spot an opportunity for improvement?Share a Content Correction