Learn More
Azlon™ Polyamide Coated Steel Bottle Carrier
Ideal for safely carrying up to six bottles or wash bottles
Brand: Azlon BGS040
94.19 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from polyamide coated steel
- Offers better heat and chemical resistance than other coatings
Specifications
Bottle Carrier | |
Polyamide-coated Steel | |
1 | |
60 mm |
Product information
File size: 2 MB
Product information
File size: 3 MB
Product Content Correction
Product Title
Azlon™ Polyamide Coated Steel Bottle Carrier > Max bottle diameter: 60mm