Learn More
Azlon™ Lid for Square Shaped HDPE Bucket
Click to view available options
Color:
Black
White
Unit Size:
Each
Description
- Lids with moulded inset handle for square shaped buckets
Specifications
Specifications
| Product Type | Lid |
| Material | HDPE |
| Autoclavable | Not Autoclavable |
| Color | Black |
| For Use With (Equipment) | Square Bucket |
| Height (Metric) | 45 mm |
| Length (Metric) | 280 mm |
| Quantity | 1 Ea. |
| Width (Metric) | 280 mm |