Learn More
Azlon™ LDPE Multi-lingual Printed Wash Bottles
Product Code. p-8001800
Click to view available options
Capacity (Metric):
250 mL
500 mL
Chemical Label:
Acetone
Deionized Water
Distilled Water
Ethanol
I. M. S.
Isopropanol
Methanol
Water
Description:
Indelibly printed in Arabic, French and English
Indelibly printed in English, French, German and Spanish
Indelibly printed in Norwegian, Danish, Swedish and English
Indelibly printed in Norwegian, Dutch, Swedish and English
Indelibly printed in Russian, Polish, German and English
Unit Size:
Pack of 5
Product Code. 12580446
Brand: Azlon™ WGW535PML
Only null left
Add to basket
Add to basket
Imprinted with Globally Harmonized Symbols, NFPA diamond, risk phrases and safety information. Azlon™ LDPE Multi-lingual Printed Wash Bottles display chemical names and key safety information in four European languages, eliminating the need for multiple wash bottles across European language speaking countries.
  • Multi-lingual wide neck wash bottles
  • Indelibly printed
  • Fitted with color-coded, one-piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
  • Available in three water and five solvent labels
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Translucent
Description Indelibly printed in English, French, German and Spanish
Includes Orange Polypropylene closure
Material LDPE
Quantity 5/PK
Temperature Range (Metric) –50 to 80°C
Closure Color Orange
Closure Included Yes
Graduated No
Mouth Wide
Shape Cylindrical
Height (Metric) 140 mm
Product Type Multi-Lingual Printed Wash Bottle
Chemical Label Ethanol
Closure Material Polypropylene
Diameter (Metric) 60 mm
Closure Type Non-venting Cap
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable