missing translation for 'onlineSavingsMsg'
Learn More

Azido-PEG3-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals

Product Code. 15416729
5 mg
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

It is suitable for confocal laser scanning microscopy. Its long PEG spacer should reduce any steric effect of the dye on the molecule to be labeled. Carboxyrhodamine 110 and its derivatives are superior alternatives to other green fluorescent dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is suitable for confocal laser scanning microscopy. Its long PEG spacer should reduce any steric effect of the dye on the molecule to be labeled. Carboxyrhodamine 110 and its derivatives are superior alternatives to other green fluorescent dyes.

Solubility
Soluble in DMSO, DMF or methanol.

Notes
Excite using the 488nm spectral line of the argon ion laser
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1536327-95-3
Molecular Formula C29H30N6O7
MDL Number MFCD22380746
Synonym Azide-Fluor 488

Specifications

Color Red
Quantity 5 mg
Solubility Information Soluble in DMSO,DMF or methanol.
Formula Weight 574.59
Physical Form Solid
Chemical Name or Material Azido-PEG3-carboxyrhodamine 110 conjugate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azido-PEG3-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.