Learn More
Tocris Bioscience™ AZ 20
Potent and selective ATR kinase inhibitor; antitumor
Brand: Tocris Bioscience™ 5198/10
293.73 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
AZ 20 is a potent and selective ATR kinase inhibitor (IC50 = 5 nM). Exhibits 7.6-fold selectivity over mTOR and selectivity over a panel of 442 kinases, including ATM kinase, PI3-K isoforms, and DNA-PK. Inhibits cell growth in cell lines with high baseline levels of replication stress. Displays antitumor effects in vivo.Specifications
>98% | |
AZ 20 | |
AZ 20 |
10 mg | |
412.51 |
For Research Use Only