Learn More
Tocris Bioscience™ AZ 20
Potent and selective ATR kinase inhibitor; antitumor
Brand:  Tocris Bioscience™ 5198/10
293.73 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18744888
  • € 331.00 / 10mg
Explore more special offers
Description
Description
AZ 20 is a potent and selective ATR kinase inhibitor (IC50 = 5 nM). Exhibits 7.6-fold selectivity over mTOR and selectivity over a panel of 442 kinases, including ATM kinase, PI3-K isoforms, and DNA-PK. Inhibits cell growth in cell lines with high baseline levels of replication stress. Displays antitumor effects in vivo.
Specifications
Specifications
>98%
AZ 20
AZ 20
10 mg
412.51
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only