Learn More
Axygen™ Tip Refill System
Axygen™ style and universal fit pipet tips in easy-to-load refill racks
Brand:  Axygen™ RFL-222-C
 View more versions of this product
Product Code. 12551288
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    1 to 200 μL
    Clear
    Universal
    Yes
    Non-sterile
    Autoclavable
    96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Axygen™ Tip Refill System > 2 Stacks of 5 x 96 Tips/Pk, 5Pk/Cs; 200 μL