Learn More
Axygen™ Polypropylene Single Well Reagent Reservoir
Durable and cost effective
Brand:  Axygen™ RES-SW8-HP-S
Product Code. 12577877
  • € 397.07 / Pack of 25
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
  • Durable and cost effective
  • Various formats for multiple applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Certified RNase-/DNase-free
Polypropylene
Reagent Reservoir
25
Flat
Clear
Sterile
25
8
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers