missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Low Profile 8-Strip PCR Tubes
GREENER_CHOICE

Product Code. 13409818 Shop All Axygen Scientific Products
Click to view available options
Color:
Clear
White
Product Type:
Thin Wall 8-Strip PCR Tube
Thin Wall 8-Strip PCR Tube with Real Time Strip Caps
Unit Size:
Pack of 125
Pack of 1250
This item is not returnable. View return policy

Product Code. 13409818

Brand: Axygen™ PCR0108LPC

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Axygen™ brand low profile 8-strip PCR tubes are ideal for low sample volume reactions.

Axygen™ Low Profile 8-Strip PCR tubes are ideal for low sample volume reactions. Using highly polished and precise molds, our tubes ensure precise thermal transfer and a leak-proof seal with a choice of strip caps. The low profile design allows reduced reaction time and evaporation due to less dead air volume. Tubes are available in white and clear with or without caps. Tubes are DNase-/RNase-free and nonpyrogenic.

 

  • Low profile design results in faster and more efficient PCR reactions
  • Ideal for 0.1 mL thermal cyclers
  • Available in white or clear
  • DNase-/RNase-free and Nonpyrogenic
  • Recommended volume range = 5 μL to 125 μL
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase and non-pyrogenic free
Color Clear
Volume (Metric) 0.1 mL
Quantity 125/Pk.
For Use With (Equipment) 0.1 mL thermal cycler for low sample volume reactions
Shape Conical
Sterility Non-sterile
Material Polypropylene
Format Strip
Product Type Thin Wall 8-Strip PCR Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.