Learn More
Axygen™ AxyMats™ Sealing Mat for 96 Well PCR Microplates
Shop All Axygen Scientific Products
Click to view available options
Material:
Silicone
missing translation for 'unitSize'
Pack of 50
Description
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Specifications
| Color | Clear |
| Material | Silicone |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| No. of Wells | 96 |
| No. per Case | 50/Cs. |
| No. per Pack | 10/Pk. |
| Product Type | Sealing Mat |
| Well Shape | Round |
| Plate Compatibility | 96-well PCR Microplate |