missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 200μL Universal Pipette Tips200μL, Non-Beveled
Designed to fit a wide variety of single and multi-channel pipettes
Brand: Axygen™ T-210-Y
This item is not returnable.
View return policy
Description
- Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
- Maxymum recovery allows complete retrieval of precious samples, saves costly reagents
- Wide bore tips of orifice size: 0.062 in. (1.52mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
- Graduation marks allow a visual confirmation of pipettor draw volume
- Presterilized tips are e-beam irradiated to prevent product degradation
- Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
Order Info
Available in bulk, racked or stack rack formats.
Specifications
1 to 200 μL | |
Yellow | |
DNase, RNase, DNA, and PCR inhibitor Free | |
Universal Fit | |
Polypropylene |
Non-sterile | |
Autoclavable | |
Universal | |
Yes |