missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Wide Bore

Product Code. 12635495 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 20000
Pack of 4800
Pack of 960
This item is not returnable. View return policy

Product Code. 12635495

Brand: Axygen™ T205WBCRS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed to fit a wide variety of single and multi-channel pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

(This is the suggested replacement for T-205-WB-C-L)

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore

Order Info

Available in bulk, racked or stack rack formats.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Genomic/Wide Orifice, Universal
For Use With (Equipment) Universal Fit
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.