Learn More
Trajan™ Autosampler Syringes With Removable Needle for Thermo Scientific GCs
Click to view available options
Needle Gauge:
23 G
26 G
Diameter (English) Outer Needle:
0.024 in.
0.18 in.
0.24 in.
Length (English) Needle:
2.24 in.
3.34 in.
Model:
0.5BR-RTC/RSH-5.7/0.63C
10R-RTC/RSH-5.7/0.47C
10R-RTC/RSH-5.7/0.63C
10R-RTC/RSH-GT-5.7/0.47C
10R-RTC/RSH-GT-5.7/0.63C
10R-RTC/RSH-GT-8.5/0.47C
10R-RTC/RSH-GT-8.5/0.63C
1BR-RTC/RSH-5.7/0.63C
5R-RTC/RSH-5.7/0.47C
5R-RTC/RSH-5.7/0.63C
5R-RTC/RSH-8.5/0.47C
Unit Size:
Each
Description
- Borosilicate glass barrel for strength and inertness.
- Stainless steel needles for inertness, strength and corrosion resistance.
- Backing behind the scale aids quick and accurate reading of the syringe scale.
- Metal back flange provides stability and resists fracture.
- Metal needle cap on syringe barrel protects and improves syringe lifetime.
- Syringes are individually tested for quality ensuring reliability and conformance.
- Unique packaging design eliminates possible contamination in the syringe.
Specifications
Specifications
| Needle Gauge | 23 G |
| Diameter (English) Outer Needle | 0.24 in. |
| Diameter (Metric) Outer Needle | 0.63 mm |
| For Use With (Application) | Gas chromatography |
| Length (English) Needle | 2.24 in. |
| Product Type | Autosampler Syringe |
| Model | 10R-RTC/RSH-5.7/0.63C |
| Quantity | Ea. |
| Volume (Metric) | 10 μL |
| Needle Point Style | Cone |
| Show More |