Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Fixed Needle for Thermo Scientific GCs
Click to view available options
Volume (Metric):
5 μL
10 μL
Needle Point Style:
Bevel
Cone
Product Type:
Fixed Needle
Removable Needle
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Thermo Scientific instruments
Specifications
Specifications
| Description | CTC/Thermo (Classic Button) Syringe |
| Needle Gauge | 23 G |
| Diameter (Metric) Outer Needle | 0.63 mm |
| Product Type | Removable Needle |
| Volume (Metric) | 10 μL |
| Needle Point Style | Cone |
| For Use With (Equipment) | Thermo Gas Chromatography |