Learn More
Trajan Scientific and Medical™ Autosampler Syringes for Agilent GCs
Product Code. 10390371
Click to view available options
Volume (Metric):
5 μL
10 μL
Needle Gauge:
23 G
23 and 26 G
26 G
Needle Point Style:
Cone
Side Hole
Unit Size:
Each
Pack of 25
Product Code. 10390371
Brand: Trajan Scientific and Medical™ 001800
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes for Agilent GCs has the knowledge and expertise to manufacture an SGE syringe.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Agilent instruments
Specifications
Needle Gauge 26 G
Diameter (Metric) Inner Needle 0.11 mm
Diameter (Metric) Outer Needle 0.47 mm
Product Type Fixed Needle Syringe
Volume (Metric) 5 μL
Needle Point Style Cone