Learn More
MedChemExpress ATP-polyamine-biotin
Click to view available options
Quantity:
1 mg
5 mg
100 mg
Unit Size:
1mg
2mg
5mg
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Biochemical Assay Reagents |
| Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Shipping Condition | Room temperature |
| CAS | 1800401-93-7 |
| Molecular Weight (g/mol) | 945.85 g/mol |
| Solubility | H2O : 6 mg/mL (6.34 mM; Need ultrasonic and warming) |
| Target | HBV |